missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine, 99%

CAS: 56-89-3 | C6H12N2O4S2 | 240.292 g/mol

Supplier:  Thermo Scientific Chemicals A137620B

 View more versions of this product

Catalog No. AAA137620B


Only null left
Explore available promotions

Description

Description

L-Cystine is used as an antioxidant, protecting tissues against radiation and pollution. It finds application in protein synthesis. It is required for utilization of vitamin B6 and is useful in healing burns and wounds. It is also is required by certain malignant cell lines in the culture medium as well as for the growth of certain micro-organisms. It is useful in the stimulation of hematopoietic system and promotes the formation of white and red blood cells. It is an active ingredient in medications used to treat dermatitis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Cystine is used as an antioxidant, protecting tissues against radiation and pollution. It finds application in protein synthesis. It is required for utilization of vitamin B6 and is useful in healing burns and wounds. It is also is required by certain malignant cell lines in the culture medium as well as for the growth of certain micro-organisms. It is useful in the stimulation of hematopoietic system and promotes the formation of white and red blood cells. It is an active ingredient in medications used to treat dermatitis.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

>240°C (decomposition)
56-89-3
C6H12N2O4S2
1728094
l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
LEVWYRKDKASIDU-IMJSIDKUSA-N
C(C(C(=O)O)N)SSCC(C(=O)O)N
240.292
67678
−219° (c=1 in 1N HCl)
99%
L-Cystine
99%
MFCD00064228
14,2782
Soluble in water.
1.68
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
1000 g
CHEBI:16283
240.3

Chemical Identifiers

56-89-3
240.292
LEVWYRKDKASIDU-IMJSIDKUSA-N
67678
(2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
C6H12N2O4S2
MFCD00064228
l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
CHEBI:16283
C(C(C(=O)O)N)SSCC(C(=O)O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-296-3

RTECSNumber : HA2690000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only