missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cysteine hydrochloride monohydrate, MP Biomedicals™
$739.13 - $739.13
Chemical Identifiers
| CAS | 4-6-7048 |
|---|---|
| Molecular Formula | C3H10ClNO3S |
| Molecular Weight (g/mol) | 175.63 |
| MDL Number | MFCD00065606 |
| InChI Key | QIJRTFXNRTXDIP-JIZZDEOASA-N |
| Synonym | l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl |
| PubChem CID | 23462 |
| IUPAC Name | (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride |
| SMILES | C(C(C(=O)O)N)S.O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10144601
|
MP Biomedicals Inc
0210144601 |
1 kg |
N/A
|
|
|||||
Chemical Identifiers
| 4-6-7048 | |
| 175.63 | |
| QIJRTFXNRTXDIP-JIZZDEOASA-N | |
| 23462 | |
| C(C(C(=O)O)N)S.O.Cl |
| C3H10ClNO3S | |
| MFCD00065606 | |
| l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl | |
| (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride |
Specifications
| 4-6-7048 | |
| MFCD00065606 | |
| QIJRTFXNRTXDIP-JIZZDEOASA-N | |
| (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride | |
| 175.63 | |
| 175.64 | |
| Crystals |
| C3H10ClNO3S | |
| l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl | |
| C(C(C(=O)O)N)S.O.Cl | |
| 1 kg | |
| 23462 | |
| Cell Culture | |
| L-Cysteine Hydrochloride Monohydrate, Precursor in the food, pharmaceutical, personal care industries |
Safety and Handling
Recommended Storage : Store at Room Temperature (15° to 30°C), store desiccated