missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine hydrochloride monohydrate, MP Biomedicals™

Catalog No. ICN10144601
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10144601 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10144601 Supplier MP Biomedicals Inc Supplier No. 0210144601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4-6-7048 | C3H10ClNO3S | 175.63 g/mol

Chemical Identifiers

CAS 4-6-7048
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
MDL Number MFCD00065606
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
Synonym l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
PubChem CID 23462
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Quantity 1 kg
Formula Weight 175.64
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Cysteine Hydrochloride Monohydrate, Precursor in the food, pharmaceutical, personal care industries
Recommended Storage Store at Room Temperature (15° to 30°C), store desiccated