missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine Hydrochloride Monohydrate, 98.5 to 101.5% (Dry Basis)

Catalog No. AAJ1403514
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1403514 25 g Plastic Bottle
AAJ1403522 100 g Plastic Bottle
AAJ14035A1 1 kg Plastic Bottle
AAJ14035A7 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ1403514 Supplier Thermo Fisher Scientific Supplier No. J1403514

CAS: 7048-04-6 | C3H10ClNO3S | 175.63 g/mol

Chemical Identifiers

CAS 4-6-7048
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
MDL Number MFCD00065606
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
Synonym l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.H2O, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
PubChem CID 23462
IUPAC Name (2 R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Melting Point ∼175°C (decomposition)
Packaging Plastic Bottle
Linear Formula HSCH2CH(NH2)COOH.HCl.H2O
Beilstein 5158059
Quantity 25 g
Formula Weight 175.63
Percent Purity 98.5 to 101.5%
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Cysteine Hydrochloride Monohydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-157-7
RTECSNumber HA2285000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only