missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine [Free Base], MP Biomedicals

Catalog No. ICN19464690
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19464690 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19464690 Supplier MP Biomedicals Inc Supplier No. 0219464690

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52-90-4 | C3H7NO2S | 121.154 g/mol

Chemical Identifiers

CAS 52-90-4
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.154
InChI Key XUJNEKJLAYXESH-REOHCLBHSA-N
Synonym l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
PubChem CID 5862
ChEBI CHEBI:17561
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid
SMILES C(C(C(=O)O)N)S

Specifications

Quantity 500 g
Formula Weight 121.2
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Cysteine [Free Base]
Recommended Storage Room temperature