missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Cysteine, Cell Culture Reagent

Catalog No. AAJ6374514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6374514 25 g
AAJ6374522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6374514 Supplier Thermo Scientific Chemicals Supplier No. J6374514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52-90-4 | C3H7NO2S | 121.154 g/mol

Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement. l-cysteine to media protects against or reverses abnormal growth & malignant transformation in aged controls. L-Cysteine is a food additive permitted for direct addition to food for human consumption.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement. l-cysteine to media protects against or reverses abnormal growth & malignant transformation in aged controls. L-Cysteine is a food additive permitted for direct addition to food for human consumption.

Solubility
Soluble in water (10 mg/ml), 1N HCl (50 mg/ml), and ethanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from air.

Chemical Identifiers

CAS 52-90-4
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.154
MDL Number MFCD00064306
InChI Key XUJNEKJLAYXESH-REOHCLBHSA-N
Synonym l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
PubChem CID 5862
ChEBI CHEBI:17561
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid
SMILES C(C(C(=O)O)N)S

Specifications

Melting Point ∽220°C (decomposition)
Color White
Beilstein 1721408
Merck Index 14,2781
Solubility Information Soluble in water (10mg/ml),1N HCl (50mg/ml),and ethanol.
Quantity 25 g
Formula Weight 121.16
Optical Rotation +7.6° (c=5 in 5N HCl)
Sensitivity Air Sensitive
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Cysteine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 200-158-2
RTECSNumber HA1600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only