missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine, 99.5%, MP Biomedicals™

Catalog No. ICN10144480
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10144480 100 g
ICN10144425 25 g
ICN10144490 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10144480 Supplier MP Biomedicals Inc Supplier No. 0210144480
Only null left

Chemical Identifiers

CAS 52-90-4
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.154
InChI Key XUJNEKJLAYXESH-REOHCLBHSA-N
Synonym l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
PubChem CID 5862
ChEBI CHEBI:17561
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid
SMILES C(C(C(=O)O)N)S

Specifications

Color White
Assay Percent Range 99.5%
Quantity 100 g
Formula Weight 121.16
Physical Form Crystalline Powder
Chemical Name or Material L-Cysteine