missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Cysteine, 98+%

Catalog No. AAA1043530
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1043530 250 g
AAA1043518 50 g
AAA104350B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1043530 Supplier Thermo Scientific Chemicals Supplier No. A1043530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L-Cysteine is used as a precursor in food, pharmaceutical and personal care products. It is actively involved in the production of flavors such as meat flavor obtained by reacting with sugars. Also used for the cleavage of disulfide bonds in the hair's keratin. It plays an important role as a key raw material for the manufacture of N-acetyl-L-cysteine, S-carboxymethyl-L-cysteine and L-cysteine base. Its derivative, N-acetyl-L-cysteine is used as a dietary supplement. It acts as an antidote to acetaminophen overdose and prevents liver failure. It protects nerves from oxidation and thereby prevents illnesses like Alzheimer's and Parkinson's. Being an essential amino acid, it used as animal feed especially for sheep, which is responsible the growth of wool.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Cysteine is used as a precursor in food, pharmaceutical and personal care products. It is actively involved in the production of flavors such as meat flavor obtained by reacting with sugars. Also used for the cleavage of disulfide bonds in the hair′s keratin. It plays an important role as a key raw material for the manufacture of N-acetyl-L-cysteine, S-carboxymethyl-L-cysteine and L-cysteine base. Its derivative, N-acetyl-L-cysteine is used as a dietary supplement. It acts as an antidote to acetaminophen overdose and prevents liver failure. It protects nerves from oxidation and thereby prevents illnesses like Alzheimer′s and Parkinson′s. Being an essential amino acid, it used as animal feed especially for sheep, which is responsible the growth of wool.

Solubility
Soluble in water, alcohol, acetic acid and aqueous ammonia. Insoluble in ether, acetone, ethyl acetate, benzene, carbon disulfide and carbon tetrachloride

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 52-90-4
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.154
MDL Number MFCD00064306
InChI Key XUJNEKJLAYXESH-REOHCLBHSA-N
Synonym l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
PubChem CID 5862
ChEBI CHEBI:17561
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid
SMILES C(C(C(=O)O)N)S

Specifications

Melting Point ∼220°C (decomposition)
Beilstein 1721408
Merck Index 14,2781
Solubility Information Soluble in water,alcohol,acetic acid and aqueous ammonia. Insoluble in ether,acetone,ethyl acetate,benzene,carbon disulfide and carbon tetrachloride
Quantity 250 g
Odor Odorless
Optical Rotation +7.9° (c=5 in 5N HCl)
Formula Weight 121.16
Sensitivity Air Sensitive
Percent Purity ≥98%
Chemical Name or Material L-Cysteine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 200-158-2
RTECSNumber HA1600000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only