missing translation for 'onlineSavingsMsg'
Learn More

L-Cyclopentylglycine, 95%

CAS: 2521-84-8 | C7H13NO2 | 143.186 g/mol

$450.15 - $1788.26

Chemical Identifiers

CAS 2521-84-8
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD03788076
InChI Key XBPKRVHTESHFAA-LURJTMIESA-N
Synonym l-cyclopentylglycine, s-2-amino-2-cyclopentylacetic acid, 2-cyclopentyl-l-glycine, h-cyclopentyl-gly-oh, l-beta-cyclopentylglycine, 2s-amino cyclopentyl ethanoic acid, h-l-cpg-oh, s-2-cyclopentylglycine, s-amino-cyclopentyl-acetic acid, 2s-2-amino-2-cyclopentylacetic acid
PubChem CID 10374564
IUPAC Name (2S)-2-amino-2-cyclopentylacetic acid
SMILES C1CCC(C1)C(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6277603
View Documents
Thermo Scientific Chemicals
H6277603
1 g
Each for $450.15
Only null left
 
AAH6277606
View Documents
Thermo Scientific Chemicals
H6277606
5 g
Each for $1,788.26
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2521-84-8
143.186
XBPKRVHTESHFAA-LURJTMIESA-N
10374564
C1CCC(C1)C(C(=O)O)N
C7H13NO2
MFCD03788076
l-cyclopentylglycine, s-2-amino-2-cyclopentylacetic acid, 2-cyclopentyl-l-glycine, h-cyclopentyl-gly-oh, l-beta-cyclopentylglycine, 2s-amino cyclopentyl ethanoic acid, h-l-cpg-oh, s-2-cyclopentylglycine, s-amino-cyclopentyl-acetic acid, 2s-2-amino-2-cyclopentylacetic acid
(2S)-2-amino-2-cyclopentylacetic acid

Specifications

2521-84-8
MFCD03788076
XBPKRVHTESHFAA-LURJTMIESA-N
(2S)-2-amino-2-cyclopentylacetic acid
143.186
143.18
L-Cyclopentylglycine
C7H13NO2
l-cyclopentylglycine, s-2-amino-2-cyclopentylacetic acid, 2-cyclopentyl-l-glycine, h-cyclopentyl-gly-oh, l-beta-cyclopentylglycine, 2s-amino cyclopentyl ethanoic acid, h-l-cpg-oh, s-2-cyclopentylglycine, s-amino-cyclopentyl-acetic acid, 2s-2-amino-2-cyclopentylacetic acid
C1CCC(C1)C(C(=O)O)N
1 g
10374564
95%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only