missing translation for 'onlineSavingsMsg'
Learn More

L-Cyclopentylglycine, 95%

Catalog No. AAH6277606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6277606 5 g
AAH6277603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6277606 Supplier Thermo Scientific Chemicals Supplier No. H6277606
Only null left

CAS: 2521-84-8 | C7H13NO2 | 143.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2521-84-8
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD03788076
InChI Key XBPKRVHTESHFAA-LURJTMIESA-N
Synonym l-cyclopentylglycine, s-2-amino-2-cyclopentylacetic acid, 2-cyclopentyl-l-glycine, h-cyclopentyl-gly-oh, l-beta-cyclopentylglycine, 2s-amino cyclopentyl ethanoic acid, h-l-cpg-oh, s-2-cyclopentylglycine, s-amino-cyclopentyl-acetic acid, 2s-2-amino-2-cyclopentylacetic acid
PubChem CID 10374564
IUPAC Name (2S)-2-amino-2-cyclopentylacetic acid
SMILES C1CCC(C1)C(C(=O)O)N

Specifications

Quantity 5 g
Formula Weight 143.18
Percent Purity 95%
Chemical Name or Material L-Cyclopentylglycine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only