missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(-)-Carvone, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 0.9580g/mL |
| Boiling Point | 227.0°C to 230.0°C |
| Flash Point | 88°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4950 to 1.5020 |
| Quantity | 100 g |
| Beilstein | 07, 157 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing. |
| EINECSNumber | 229-352-5 |
| RTECSNumber | OS8650000 |
| TSCA | TSCA |
RUO – Research Use Only