missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Carnitine Hydrochloride 98.0+%, TCI America™
$76.43 - $240.46
Chemical Identifiers
| CAS | 6645-46-1 |
|---|---|
| Molecular Formula | C7H16ClNO3 |
| Molecular Weight (g/mol) | 197.66 |
| MDL Number | MFCD00066100 |
| InChI Key | JXXCENBLGFBQJM-FYZOBXCZSA-N |
| Synonym | r-3-carboxy-2-hydroxy-n,n,n-trimethylpropan-1-aminium chloride, l-carnitine hcl, unii-j3y5e6iks3, l-carnitine hydrochloride, levocarnitine chloride, j3y5e6iks3, r-carnitine hydrochloride, carnitine l-form hydrochloride, l-3-carboxy-2-hydroxypropyl trimethylammonium chloride |
| PubChem CID | 656657 |
| IUPAC Name | [(2R)-3-carboxy-2-hydroxypropyl]trimethylazanium chloride |
| SMILES | [Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O |
Chemical Identifiers
| 6645-46-1 | |
| 197.66 | |
| JXXCENBLGFBQJM-FYZOBXCZSA-N | |
| 656657 | |
| [Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O |
| C7H16ClNO3 | |
| MFCD00066100 | |
| r-3-carboxy-2-hydroxy-n,n,n-trimethylpropan-1-aminium chloride, l-carnitine hcl, unii-j3y5e6iks3, l-carnitine hydrochloride, levocarnitine chloride, j3y5e6iks3, r-carnitine hydrochloride, carnitine l-form hydrochloride, l-3-carboxy-2-hydroxypropyl trimethylammonium chloride | |
| [(2R)-3-carboxy-2-hydroxypropyl]trimethylazanium chloride |
Specifications
| 6645-46-1 | |
| White | |
| MFCD00066100 | |
| r-3-carboxy-2-hydroxy-n,n,n-trimethylpropan-1-aminium chloride, l-carnitine hcl, unii-j3y5e6iks3, l-carnitine hydrochloride, levocarnitine chloride, j3y5e6iks3, r-carnitine hydrochloride, carnitine l-form hydrochloride, l-3-carboxy-2-hydroxypropyl trimethylammonium chloride | |
| [Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O | |
| 197.66 | |
| 197.66 | |
| Crystalline Powder |
| 139°C | |
| C7H16ClNO3 | |
| 5 g | |
| JXXCENBLGFBQJM-FYZOBXCZSA-N | |
| [(2R)-3-carboxy-2-hydroxypropyl]trimethylazanium chloride | |
| 656657 | |
| ≥98.0% (T) | |
| L-Carnitine Hydrochloride |
Safety and Handling
EINECSNumber : (9)-0253
RTECSNumber : BP2979100
TSCA : No