missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Carnitine, 99+%

Catalog No. AC241040100
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC241040100 10 g Glass bottle
AC241040010 1 g Glass bottle
AC241040500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC241040100 Supplier Thermo Scientific Chemicals Supplier No. 241040100
Only null left

CAS: 541-15-1 | C7H15NO3 | 161.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 541-15-1
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.2
MDL Number MFCD00038747
InChI Key PHIQHXFUZVPYII-ZCFIWIBFSA-N
Synonym l-carnitine, levocarnitine, vitamin bt, r-carnitine, carnitor, --carnitine, carnitine, --l-carnitine, karnitin, l---carnitine
PubChem CID 10917
ChEBI CHEBI:16347
IUPAC Name (3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate
SMILES C[N+](C)(C)CC(CC(=O)[O-])O

Specifications

Melting Point 208°C to 212°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (CH3)3N+CH2CH(OH)CH2COO-
Quantity 10 g
Beilstein 04, IV, 3185
Merck Index 15, 1849
Specific Rotation -31
Solubility Information (5% in water) Clear colorless
Formula Weight 161.2
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(-)-Carnitine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-768-

RUO – Research Use Only