Learn More
L-Carnitine, 98+%
CAS: 541-15-1 | C7H15NO3 | 161.201 g/mol
Supplier: Thermo Scientific Chemicals A1761818
Description
L-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.
Solubility
Soluble in water, pyridine, dimethyl sulfoxide, methanol and ethanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
Specifications
| L-Carnitine | |
| ∼190°C (decomposition) | |
| MFCD00038747 | |
| 4292315 | |
| 14,1849 | |
| Soluble in water,pyridine,dimethyl sulfoxide,methanol and ethanol. | |
| C[N+](C)(C)CC(CC(=O)[O-])O | |
| 161.201 | |
| CHEBI:16347 | |
| ≥99% |
| 541-15-1 | |
| C7H15NO3 | |
| 50 g | |
| Hygroscopic | |
| l-carnitine, levocarnitine, vitamin bt, r-carnitine, carnitor, --carnitine, carnitine, --l-carnitine, karnitin, l---carnitine | |
| PHIQHXFUZVPYII-ZCFIWIBFSA-N | |
| (3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate | |
| 10917 | |
| 161.2 |
Chemical Identifiers
| 541-15-1 | |
| 161.201 | |
| PHIQHXFUZVPYII-ZCFIWIBFSA-N | |
| 10917 | |
| (3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate |
| C7H15NO3 | |
| MFCD00038747 | |
| l-carnitine, levocarnitine, vitamin bt, r-carnitine, carnitor, --carnitine, carnitine, --l-carnitine, karnitin, l---carnitine | |
| CHEBI:16347 | |
| C[N+](C)(C)CC(CC(=O)[O-])O |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-768-0
RTECSNumber : BP2980000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only