missing translation for 'onlineSavingsMsg'
Learn More

L-Carnitine, 98+%

CAS: 541-15-1 | C7H15NO3 | 161.201 g/mol

Supplier:  Thermo Scientific Chemicals A1761809

 View more versions of this product

Catalog No. AAA1761809


Only null left
Explore available promotions

Description

Description

L-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.

Solubility
Soluble in water, pyridine, dimethyl sulfoxide, methanol and ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
Specifications

Specifications

L-Carnitine
∼190°C (decomposition)
MFCD00038747
4292315
14,1849
Soluble in water,pyridine,dimethyl sulfoxide,methanol and ethanol.
C[N+](C)(C)CC(CC(=O)[O-])O
161.201
CHEBI:16347
≥99%
541-15-1
C7H15NO3
10 g
Hygroscopic
l-carnitine, levocarnitine, vitamin bt, r-carnitine, carnitor, --carnitine, carnitine, --l-carnitine, karnitin, l---carnitine
PHIQHXFUZVPYII-ZCFIWIBFSA-N
(3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate
10917
161.2

Chemical Identifiers

541-15-1
161.201
PHIQHXFUZVPYII-ZCFIWIBFSA-N
10917
(3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate
C7H15NO3
MFCD00038747
l-carnitine, levocarnitine, vitamin bt, r-carnitine, carnitor, --carnitine, carnitine, --l-carnitine, karnitin, l---carnitine
CHEBI:16347
C[N+](C)(C)CC(CC(=O)[O-])O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-768-0

RTECSNumber : BP2980000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only