missing translation for 'onlineSavingsMsg'
Learn More

L-Buthionine-(S,R)-sulfoximine, 99%

Catalog No. AC235520010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC235520010 1 g Glass bottle
AC235520050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC235520010 Supplier Thermo Scientific Chemicals Supplier No. 235520010

CAS: 83730-53-4 | C8H18N2O3S | 222.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83730-53-4
Molecular Formula C8H18N2O3S
Molecular Weight (g/mol) 222.31
MDL Number MFCD00067000
InChI Key KJQFBVYMGADDTQ-CVSPRKDYSA-N
Synonym l-buthionine-sulfoximine, l-buthionine sulfoximine, 2s-2-amino-4-butylsulfonimidoyl butanoic acid, l-buthionine-s,r-sulfoximine, l-buthionine s,r-sulfoximine, l-bso, butanoic acid, 2-amino-4-s-butylsulfonimidoyl-, 2s, l-buthionine sr-sulfoximine, buthionine-s,r-sulfoximine, l, l-buthionine-s,r-sulphoximine
PubChem CID 119565
IUPAC Name (2S)-2-amino-4-(butylsulfonimidoyl)butanoic acid
SMILES CCCCS(=N)(=O)CCC(C(=O)O)N

Specifications

Melting Point 234.0°C to 235.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOOCCH(NH2)CH2CH2S(O)(=NH)(CH2)3CH3
Quantity 1 g
Specific Rotation + 33.00
Solubility Information Solubility in water: soluble
Formula Weight 222.31
Percent Purity 99%
Physical Form Fine Powder
Chemical Name or Material L-Buthionine-(S, R)-sulfoximine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only