missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-Bornyl acetate, 95%
CAS: 5655-61-8 | C12H20O2 | 196.29 g/mol
$45.18 - $344.23
Chemical Identifiers
| CAS | 5655-61-8 |
|---|---|
| Molecular Formula | C12H20O2 |
| Molecular Weight (g/mol) | 196.29 |
| MDL Number | MFCD00867808,MFCD00135942,MFCD00135942 |
| InChI Key | KGEKLUUHTZCSIP-HOSYDEDBSA-N |
| Synonym | bornyl acetate, -bornyl acetate |
| PubChem CID | 44630108 |
| SMILES | CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2295714
|
Thermo Scientific Chemicals
B2295714 |
25 g |
Each for $45.18
|
|
|||||
|
AAB2295722
|
Thermo Scientific Chemicals
B2295722 |
100 g |
Each for $108.74
|
|
|||||
|
AAB2295736
|
Thermo Scientific Chemicals
B2295736 |
500 g |
Each for $344.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5655-61-8 | |
| 196.29 | |
| KGEKLUUHTZCSIP-HOSYDEDBSA-N | |
| 44630108 |
| C12H20O2 | |
| MFCD00867808,MFCD00135942,MFCD00135942 | |
| bornyl acetate, -bornyl acetate | |
| CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C |
Specifications
| 5655-61-8 | |
| 223°C to 224°C | |
| C12H20O2 | |
| MFCD00867808,MFCD00135942,MFCD00135942 | |
| 2502036 | |
| bornyl acetate, -bornyl acetate | |
| CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C | |
| 196.29 | |
| 196.29 | |
| L-(-)-Bornyl acetate |
| 0.98 | |
| 84°C (184°F) | |
| 1.463 | |
| 25 g | |
| 14,1339 | |
| KGEKLUUHTZCSIP-HOSYDEDBSA-N | |
| (1S,2R,4S)-1,7,7-trimethylbicyclo[2.2.1]heptan-2-yl acetate | |
| 44630108 | |
| 95% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 227-101-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only