missing translation for 'onlineSavingsMsg'
Learn More

L-Bornyl acetate, 95%

Catalog No. AC106555000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC106555000 500 g
AC106550050 5 g
AC106551000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC106555000 Supplier Thermo Scientific Chemicals Supplier No. 106555000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5655-61-8 | C12H20O2 | 196.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5655-61-8
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00867808,MFCD00135942,MFCD00135942
InChI Key KGEKLUUHTZCSIP-HOSYDEDBSA-N
Synonym bornyl acetate, -bornyl acetate
PubChem CID 44630108
IUPAC Name [(1R,3S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
SMILES CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C

Specifications

Color Colorless
Density 0.9875g/mL
Boiling Point 223°C to 224°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (as C12H20O2) Total esters (GC)
Packaging Glass bottle
Refractive Index 1.4620 to 1.465
Quantity 500 g
Beilstein 06,82
Merck Index 15,1341
Specific Gravity 0.9875
Specific Rotation -38
Solubility Information Solubility in water: .. Other solubilities: miscible with 95% alcohol and ether
Formula Weight 196.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material L-Bornyl acetate, 95%
Show More Show Less
EINECSNumber 227-101-4
TSCA TSCA

RUO – Research Use Only