missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Borneol, 97%

Catalog No. AC160655000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160655000 500 g Plastic bottle
AC160650250 25 g Plastic bottle
AC160651000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC160655000 Supplier Thermo Scientific Chemicals Supplier No. 160655000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 464-45-9 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 464-45-9
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00003759,MFCD00066426,MFCD00066427
InChI Key DTGKSKDOIYIVQL-NQMVMOMDSA-N
Synonym --borneol, borneol, l-borneol, 1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol, linderol, 1s-endo---borneol, borneo camphor, bornyl alcohol, sumatra camphor, dl-borneol
PubChem CID 1201518
ChEBI CHEBI:15394
IUPAC Name (1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
SMILES CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2

Specifications

Melting Point 204°C to 208°C
Color White to Yellow
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 06,72
Merck Index 15,134
Specific Rotation -36.2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 154.25
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(-)-Borneol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 207-353-1
RTECSNumber DT5095000
TSCA TSCA

RUO – Research Use Only