missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Borneol, 97%

CAS: 464-45-9 | C10H18O | 154.25 g/mol

Supplier:  Thermo Scientific Chemicals 160651000

Quantity 100 g
Packaging Plastic bottle

 View more versions of this product

Catalog No. AC160651000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

464-45-9
154.25
DTGKSKDOIYIVQL-NQMVMOMDSA-N
1201518
(1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
C10H18O
MFCD00003759,MFCD00066426,MFCD00066427
--borneol, borneol, l-borneol, 1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol, linderol, 1s-endo---borneol, borneo camphor, bornyl alcohol, sumatra camphor, dl-borneol
CHEBI:15394
CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2

Specifications

L(-)-Borneol
464-45-9
White to Yellow
Authentic
Plastic bottle
100 g
06,72
-36.2
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
CC1(C)[C@@H]2CC[C@]1(C)[C@H](O)C2
154.25
CHEBI:15394
97%
98%
204°C to 208°C
65°C
96% min. (GC)
C10H18O
MFCD00003759,MFCD00066426,MFCD00066427
15,134
--borneol, borneol, l-borneol, 1s,2r,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-ol, linderol, 1s-endo---borneol, borneo camphor, bornyl alcohol, sumatra camphor, dl-borneol
DTGKSKDOIYIVQL-NQMVMOMDSA-N
(1S,3R,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-ol
1201518
154.25
Crystalline Powder or Crystals
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Danger

EINECSNumber : 207-353-1

RTECSNumber : DT5095000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions