missing translation for 'onlineSavingsMsg'
Learn More

L-beta-Prolinol, 95%

CAS: 110013-19-9 | C5H11NO | 101.149 g/mol

$183.59 - $626.68

Chemical Identifiers

CAS 110013-19-9
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD09260722
InChI Key QOTUIIJRVXKSJU-YFKPBYRVSA-N
Synonym s-pyrrolidin-3-ylmethanol, 3s-pyrrolidin-3-ylmethanol, l-beta-prolinol, s-3-pyrrolidin-methanol, 3-pyrrolidinemethanol, 3s, 3-pyrrolidinemethanol, s, 3s-pyrrolidin-3-yl methanol, s-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine
PubChem CID 7446911
IUPAC Name [(3S)-pyrrolidin-3-yl]methanol
SMILES C1CNCC1CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH57199MD
View Documents
Thermo Scientific Chemicals
H57199MD
250 mg
Each for $183.59
Only null left
 
AAH5719903
View Documents
Thermo Scientific Chemicals
H5719903
1 g
Each for $626.68
Only null left
 
Description

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water

Notes
Hygroscopic. Store at room temperature.
Specifications

Chemical Identifiers

110013-19-9
101.149
QOTUIIJRVXKSJU-YFKPBYRVSA-N
7446911
C1CNCC1CO
C5H11NO
MFCD09260722
s-pyrrolidin-3-ylmethanol, 3s-pyrrolidin-3-ylmethanol, l-beta-prolinol, s-3-pyrrolidin-methanol, 3-pyrrolidinemethanol, 3s, 3-pyrrolidinemethanol, s, 3s-pyrrolidin-3-yl methanol, s-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine
[(3S)-pyrrolidin-3-yl]methanol

Specifications

110013-19-9
174°C to 176°C
Hygroscopic
MFCD09260722
Slightly soluble in water
C1CNCC1CO
101.149
101.15
L-beta-Prolinol
0.979
250 mg
C5H11NO
s-pyrrolidin-3-ylmethanol, 3s-pyrrolidin-3-ylmethanol, l-beta-prolinol, s-3-pyrrolidin-methanol, 3-pyrrolidinemethanol, 3s, 3-pyrrolidinemethanol, s, 3s-pyrrolidin-3-yl methanol, s-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine
QOTUIIJRVXKSJU-YFKPBYRVSA-N
[(3S)-pyrrolidin-3-yl]methanol
7446911
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

RUO – Research Use Only