Learn More
L-beta-Prolinol, 95%
CAS: 110013-19-9 | C5H11NO | 101.149 g/mol
Supplier: Thermo Scientific Chemicals H5719903
| Quantity | 1 g |
|---|
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water
Notes
Hygroscopic. Store at room temperature.
Chemical Identifiers
| 110013-19-9 | |
| 101.149 | |
| QOTUIIJRVXKSJU-YFKPBYRVSA-N | |
| 7446911 | |
| C1CNCC1CO |
| C5H11NO | |
| MFCD09260722 | |
| s-pyrrolidin-3-ylmethanol, 3s-pyrrolidin-3-ylmethanol, l-beta-prolinol, s-3-pyrrolidin-methanol, 3-pyrrolidinemethanol, 3s, 3-pyrrolidinemethanol, s, 3s-pyrrolidin-3-yl methanol, s-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine | |
| [(3S)-pyrrolidin-3-yl]methanol |
Specifications
| L-beta-Prolinol | |
| 0.979 | |
| 1 g | |
| C5H11NO | |
| s-pyrrolidin-3-ylmethanol, 3s-pyrrolidin-3-ylmethanol, l-beta-prolinol, s-3-pyrrolidin-methanol, 3-pyrrolidinemethanol, 3s, 3-pyrrolidinemethanol, s, 3s-pyrrolidin-3-yl methanol, s-3-pyrrolidinemethanol, s-3-hydroxymethyl pyrrolidine | |
| QOTUIIJRVXKSJU-YFKPBYRVSA-N | |
| [(3S)-pyrrolidin-3-yl]methanol | |
| 7446911 | |
| 95% |
| 110013-19-9 | |
| 174°C to 176°C | |
| Hygroscopic | |
| MFCD09260722 | |
| Slightly soluble in water | |
| C1CNCC1CO | |
| 101.149 | |
| 101.15 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold; Store under Argon