missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-beta-Proline, 98+%
CAS: 72580-53-1 | C5H9NO2 | 115.132 g/mol
$154.99 - $437.79
Chemical Identifiers
| CAS | 72580-53-1 |
|---|---|
| Molecular Formula | C5H9NO2 |
| Molecular Weight (g/mol) | 115.132 |
| MDL Number | MFCD06738463 |
| InChI Key | JAEIBKXSIXOLOL-BYPYZUCNSA-N |
| Synonym | s-pyrrolidine-3-carboxylic acid, pubchem11126, l-beta-proline, l-beta-proline, 3s-3-pyrrolidinecarboxylic acid, 3-pyrrolidinecarboxylic acid, 3s, s-pyrrolidine-3-carboxylicacid, s-beta-proline, s-beta-proline, a-proline, a-proline |
| PubChem CID | 1501969 |
| IUPAC Name | (3S)-pyrrolidine-3-carboxylic acid |
| SMILES | C1CNCC1C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH57895MD
|
Thermo Scientific Chemicals
H57895MD |
250 mg |
Each for $154.99
|
|
|||||
|
AAH5789503
|
Thermo Scientific Chemicals
H5789503 |
1 g |
Each for $437.79
|
|
|||||
Description
It is an agrochemical and pharmacetical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an agrochemical and pharmacetical intermediate.
Solubility
Slightly soluble in water.
Notes
Recommended storage temperature: 2 - 8°C. Incompatible with strong oxidizing agents, acids.
Chemical Identifiers
| 72580-53-1 | |
| 115.132 | |
| JAEIBKXSIXOLOL-BYPYZUCNSA-N | |
| 1501969 | |
| C1CNCC1C(=O)O |
| C5H9NO2 | |
| MFCD06738463 | |
| s-pyrrolidine-3-carboxylic acid, pubchem11126, l-beta-proline, l-beta-proline, 3s-3-pyrrolidinecarboxylic acid, 3-pyrrolidinecarboxylic acid, 3s, s-pyrrolidine-3-carboxylicacid, s-beta-proline, s-beta-proline, a-proline, a-proline | |
| (3S)-pyrrolidine-3-carboxylic acid |
Specifications
| 185°C to 191°C | |
| C5H9NO2 | |
| 3537293 | |
| Slightly soluble in water. | |
| C1CNCC1C(=O)O | |
| 250 mg | |
| 1501969 | |
| 115.13 | |
| L-beta-Proline |
| 72580-53-1 | |
| MFCD06738463 | |
| s-pyrrolidine-3-carboxylic acid, pubchem11126, l-beta-proline, l-beta-proline, 3s-3-pyrrolidinecarboxylic acid, 3-pyrrolidinecarboxylic acid, 3s, s-pyrrolidine-3-carboxylicacid, s-beta-proline, s-beta-proline, a-proline, a-proline | |
| JAEIBKXSIXOLOL-BYPYZUCNSA-N | |
| (3S)-pyrrolidine-3-carboxylic acid | |
| 115.132 | |
| +18° (c=2 in Water) | |
| ≥98% |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only