missing translation for 'onlineSavingsMsg'
Learn More

L-beta-Homoglutamic acid hydrochloride, 97%, Thermo Scientific Chemicals

Catalog No. p-7056451
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6537003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6537003 Supplier Thermo Scientific Chemicals Supplier No. H6537003

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61884-74-0
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.615
MDL Number MFCD01862872
InChI Key XMUKSNPNAOIQKU-WCCKRBBISA-N
Synonym s-3-aminohexanedioic acid hydrochloride, l-beta-homoglutamic acid hydrochloride, h-beta-hoglu-oh.hcl, 3s-3-aminohexanedioic acid hydrochloride, h-beta-homoglu-ohcl, h-hoglu-oh.hcl, h-?-hoglu-oh.hcl, h-beta-homo-glu-oh hcl, l-beta-homoglutamic acidhcl, l-beta-homoglutamic acid-hcl
PubChem CID 2761515
IUPAC Name (3S)-3-aminohexanedioic acid;hydrochloride
SMILES C(CC(=O)O)C(CC(=O)O)N.Cl

Specifications

Color White
Beilstein 6584879
Quantity 1 g
Formula Weight 197.62
Percent Purity 97%
Physical Form Solid
Chemical Name or Material L-beta-Homoglutamic acid hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only