missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Aspartyl-L-phenylalanine methyl ester, 98%

Catalog No. AC228650010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228650010 1 g Glass Bottle
AC228650050 5 g Glass Bottle
AC228650250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC228650010 Supplier Thermo Scientific Chemicals Supplier No. 228650010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 22839-47-0 | C14H18N2O5 | 294.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22839-47-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00002724
InChI Key IAOZJIPTCAWIRG-QWRGUYRKSA-N
Synonym aspartame, nutrasweet, asp-phe-ome, asp-phe methyl ester, aspartam, aspartamum, aspartamo, l-aspartyl-l-phenylalanine methyl ester, aspartylphenylalanine methyl ester, dipeptide sweetener
PubChem CID 134601
ChEBI CHEBI:2877
IUPAC Name (3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid
SMILES COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O

Specifications

Melting Point 248.0°C to 250.0°C
Color White
Infrared Spectrum Authentic
Loss on Drying 4.5% max. (105°C, 4 hrs)
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HO2CCH2CH(NH2)CONHCH(CH2C6H5)CO2CH3
Specific Rotation + 12.50
Transmittance 95% min. (c=1, 2N HCl, 430nm) 1cm cell
Merck Index 15, 829
Solubility Information Solubility in water: sparingly soluble.
Quantity 1 g
Formula Weight 294.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L-Aspartyl-L-phenylalanine methyl ester
Show More Show Less
EINECSNumber 245-261-3

RUO – Research Use Only