missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic Acid 99.0+%, TCI America™

Catalog No. A054625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A054625G 25 g
A0546500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A054625G Supplier TCI America Supplier No. A054625G

Chemical Identifiers

CAS 56-84-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00002616
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid
PubChem CID 5960
ChEBI CHEBI:17053
IUPAC Name 2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 133.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Aspartic Acid
EINECSNumber (2)-1305
RTECSNumber CI9098500
TSCA Yes