missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic Acid, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. p-7156945
Change view
Click to view available options
Quantity:
1 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603735 1 kg Poly Pail
18603736 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18603735 Supplier Spectrum Chemical Mfg Cor Supplier No. A13761KG
Only null left

A1376, 56-84-8, C4H7NO4

Spectrum™ Chemical L-Aspartic Acid, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-84-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00002616
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
IUPAC Name 2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Lead (Pb) 5 mg/kg
Loss on Drying 0.25%
Packaging Poly Pail
Specific Rotation +24.5° to +26.0°
Quantity 1 kg
Percent Purity 98.5 to 101.5%
Grade FCC