missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid dibenzyl ester p-toluenesulfonate, 95%, Thermo Scientific Chemicals

Catalog No. p-7056604
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6562314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6562314 Supplier Thermo Scientific Chemicals Supplier No. H6562314

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2886-33-1
Molecular Formula C25H27NO7S
Molecular Weight (g/mol) 485.55
MDL Number MFCD00065188
InChI Key HLMUYZYLPUHSNV-UHFFFAOYNA-N
Synonym h-asp obzl-obzl.tosoh, h-asp obzl-obzl tosoh, l-aspartic acid dibenzyl ester tosylate, s-dibenzyl 2-aminosuccinate 4-methylbenzenesulfonate, l-aspartic acid dibenzyl ester p-toluenesulfonate salt, l-aspartic acid dibenzyl ester 4-toluenesulfonate, h-asp obzl-obzl inverted exclamation mark currencytosoh, h-asp obzl-obzl.tos, l-aspartic acid dibenzyl ester p-toluenesulfonate, 1,4-dibenzyl 2s-2-aminobutanedioate; para-toluene sulfonate
PubChem CID 2808531
IUPAC Name 1,4-dibenzyl 2-aminobutanedioate; 4-methylbenzene-1-sulfonic acid
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.NC(CC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 157°C to 160°C
Color Brown
Beilstein 3647510
Quantity 25 g
Formula Weight 485.55
Percent Purity 95%
Physical Form Powder
Chemical Name or Material L-Aspartic acid dibenzyl ester p-toluenesulfonate
EINECSNumber 220-746-2
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only