Learn More
L-Aspartic acid, 99%, low metals content
CAS: 56-84-8 | C4H7NO4 | 133.10 g/mol
$70.51 - $179.32
Chemical Identifiers
| CAS | 56-84-8 |
|---|---|
| Molecular Formula | C4H7NO4 |
| Molecular Weight (g/mol) | 133.10 |
| MDL Number | MFCD00002616 |
| InChI Key | CKLJMWTZIZZHCS-UHFFFAOYNA-N |
| Synonym | l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid |
| PubChem CID | 5960 |
| ChEBI | CHEBI:17053 |
| IUPAC Name | (2S)-2-aminobutanedioic acid |
| SMILES | NC(CC(O)=O)C(O)=O |
Description
L-Aspartic acid is used as a component of parenteral and enteral nutrition and as a pharmaceutical ingredient. it is used for cell culture and in manufacturing processes. It is widely utilized for mineral supplementation in the salt form.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 56-84-8 | |
| 133.10 | |
| CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
| 5960 | |
| (2S)-2-aminobutanedioic acid |
| C4H7NO4 | |
| MFCD00002616 | |
| l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid | |
| CHEBI:17053 | |
| NC(CC(O)=O)C(O)=O |
Specifications
| >300°C (decomposition) | |
| C4H7NO4 | |
| 1723530 | |
| l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid | |
| CKLJMWTZIZZHCS-UHFFFAOYNA-N | |
| NC(CC(O)=O)C(O)=O | |
| 133.10 | |
| 5960 | |
| Odorless | |
| 99% | |
| L-Aspartic acid, Low metals content |
| 56-84-8 | |
| MFCD00002616 | |
| 14,840 | |
| Soluble in water(4.5g/L). | |
| 1.66 g/mL | |
| (2S)-2-aminobutanedioic acid | |
| 100 g | |
| CHEBI:17053 | |
| 133.1 | |
| Powder |
Safety and Handling
EINECSNumber : 200-291-6
RTECSNumber : CI9098500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only