missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride, 98%

Catalog No. AAH6325703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6325703 1 g
AAH6325706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6325703 Supplier Thermo Scientific Chemicals Supplier No. H6325703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2673-19-0 | C9H18ClNO4 | 239.696 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2673-19-0
Molecular Formula C9H18ClNO4
Molecular Weight (g/mol) 239.696
MDL Number MFCD00153427
InChI Key SFYKWYAIJZEDNG-RGMNGODLSA-N
Synonym h-asp otbu-ome.hcl, h-asp otbu-ome hcl, l-aspartic acid 4-tert-butyl-1-methyl ester hydrochloride, h-asp otbu-ome hydrochloride, h-asp obut-ome hcl, s-4-tert-butyl 1-methyl 2-aminosuccinate hydrochloride, 4-tert-butyl 1-methyl 2s-2-aminobutanedioate hydrochloride, h-asp otbu-ome, h-asp otbu-omecl, h-asp o-tbu-ome
PubChem CID 13970777
IUPAC Name 4-O-tert-butyl 1-O-methyl (2S)-2-aminobutanedioate;hydrochloride
SMILES CC(C)(C)OC(=O)CC(C(=O)OC)N.Cl

Specifications

Beilstein 6005591
Quantity 1 g
Formula Weight 239.7
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Aspartic acid 4-tert-butyl 1-methyl ester hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only