missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine tert-butyl ester hydrochloride, 95%

Catalog No. p-7056706
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH65792MD 250 mg
AAH6579203 1 g
AAH6579206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH65792MD Supplier Thermo Scientific Chemicals Supplier No. H65792MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 63094-81-5 | C8H17ClN2O3 | 224.685 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63094-81-5
Molecular Formula C8H17ClN2O3
Molecular Weight (g/mol) 224.685
MDL Number MFCD00039079
InChI Key RXNKCUXXNGWROA-JEDNCBNOSA-N
Synonym h-asn-otbu hcl, l-asparagine tert-butyl ester hydrochloride, h-asn-otbu.hcl, tert-butyl l-asparaginate-hydrogen chloride 1/1, l-asparagine, 1,1-dimethylethyl ester, monohydrochloride, tert-butyl 2s-2-amino-3-carbamoylpropanoate hydrochloride, h-asn-otbucl, h-asn-otbu?cl, h-asn-otbuhcl, h-asn-otbu hydrochloride
PubChem CID 16218498
IUPAC Name tert-butyl (2S)-2,4-diamino-4-oxobutanoate;hydrochloride
SMILES CC(C)(C)OC(=O)C(CC(=O)N)N.Cl

Specifications

Color White
Beilstein 6585123
Quantity 250 mg
Formula Weight 224.69
Percent Purity 95%
Physical Form Powder
Chemical Name or Material L-Asparagine tert-butyl ester hydrochloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only