missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Asparagine Monohydrate 99.0+%, TCI America™
$142.46 - $797.02
Chemical Identifiers
| CAS | 5794-13-8 |
|---|---|
| Molecular Formula | C4H10N2O4 |
| Molecular Weight (g/mol) | 150.134 |
| MDL Number | MFCD00151038 |
| InChI Key | RBMGJIZCEWRQES-DKWTVANSSA-N |
| Synonym | l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521 |
| PubChem CID | 170358 |
| IUPAC Name | (2S)-2,4-diamino-4-oxobutanoic acid;hydrate |
| SMILES | C(C(C(=O)O)N)C(=O)N.O |
Chemical Identifiers
| 5794-13-8 | |
| 150.134 | |
| RBMGJIZCEWRQES-DKWTVANSSA-N | |
| 170358 | |
| C(C(C(=O)O)N)C(=O)N.O |
| C4H10N2O4 | |
| MFCD00151038 | |
| l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521 | |
| (2S)-2,4-diamino-4-oxobutanoic acid;hydrate |
Specifications
| 235°C | |
| White | |
| MFCD00151038 | |
| RBMGJIZCEWRQES-DKWTVANSSA-N | |
| (2S)-2,4-diamino-4-oxobutanoic acid;hydrate | |
| 150.134 | |
| 132.12 | |
| Crystalline Powder |
| 5794-13-8 | |
| C4H10N2O4 | |
| l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521 | |
| C(C(C(=O)O)N)C(=O)N.O | |
| 25 g | |
| 170358 | |
| ≥99.0% (HPLC,T) | |
| L-Asparagine Monohydrate |
Safety and Handling
EINECSNumber : (2)-2383
TSCA : Yes