missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine Monohydrate 99.0+%, TCI America™

Catalog No. A054225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A054225G 25 g
A0542250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A054225G Supplier TCI America Supplier No. A054225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol

Chemical Identifiers

CAS 5794-13-8
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
MDL Number MFCD00151038
InChI Key RBMGJIZCEWRQES-DKWTVANSSA-N
Synonym l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521
PubChem CID 170358
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES C(C(C(=O)O)N)C(=O)N.O

Specifications

Melting Point 235°C
Color White
Quantity 25 g
Formula Weight 132.12
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Asparagine Monohydrate
EINECSNumber (2)-2383
TSCA Yes