missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Asparagine monohydrate, Cell Culture Reagent
CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol
$171.07 - $1135.10
Chemical Identifiers
| CAS | 5794-13-8 |
|---|---|
| Molecular Formula | C4H10N2O4 |
| Molecular Weight (g/mol) | 150.134 |
| MDL Number | MFCD00151038 |
| InChI Key | RBMGJIZCEWRQES-DKWTVANSSA-N |
| Synonym | l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521 |
| PubChem CID | 170358 |
| IUPAC Name | (2S)-2,4-diamino-4-oxobutanoic acid;hydrate |
| SMILES | C(C(C(=O)O)N)C(=O)N.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6286922
|
Thermo Scientific Chemicals
J6286922 |
100 g |
Each for $171.07
|
|
|||||
|
AAJ6286936
|
Thermo Scientific Chemicals
J6286936 |
500 g |
Each for $700.82
|
|
|||||
|
AAJ62869A1
|
Thermo Scientific Chemicals
J62869A1 |
1 kg |
Each for $1,135.10
|
|
|||||
Description
It is used as an isotopically labeled biochemical for proteomics research. It is suitable for all food, beverage and nutritional supplement applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an isotopically labeled biochemical for proteomics research. It is suitable for all food, beverage and nutritional supplement applications.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
| 5794-13-8 | |
| 150.134 | |
| RBMGJIZCEWRQES-DKWTVANSSA-N | |
| 170358 | |
| C(C(C(=O)O)N)C(=O)N.O |
| C4H10N2O4 | |
| MFCD00151038 | |
| l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521 | |
| (2S)-2,4-diamino-4-oxobutanoic acid;hydrate |
Specifications
| 215°C to 217°C | |
| White | |
| MFCD00151038 | |
| l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521 | |
| RBMGJIZCEWRQES-DKWTVANSSA-N | |
| (2S)-2,4-diamino-4-oxobutanoic acid;hydrate | |
| 150.134 | |
| 150.14 (132.12 Anhydrous) | |
| Powder |
| 5794-13-8 | |
| C4H10N2O4 | |
| 1723527 | |
| Soluble in water. | |
| C(C(C(=O)O)N)C(=O)N.O | |
| 100 g | |
| 170358 | |
| Cell Culture Reagent | |
| L-Asparagine monohydrate |
Safety and Handling
EINECSNumber : 200-735-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only