missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Asparagine monohydrate, Cell Culture Reagent

Catalog No. AAJ62869A1
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62869A1 1 kg
AAJ6286922 100 g
AAJ6286936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62869A1 Supplier Thermo Scientific Chemicals Supplier No. J62869A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol

It is used as an isotopically labeled biochemical for proteomics research. It is suitable for all food, beverage and nutritional supplement applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an isotopically labeled biochemical for proteomics research. It is suitable for all food, beverage and nutritional supplement applications.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 5794-13-8
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
MDL Number MFCD00151038
InChI Key RBMGJIZCEWRQES-DKWTVANSSA-N
Synonym l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521
PubChem CID 170358
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES C(C(C(=O)O)N)C(=O)N.O

Specifications

Melting Point 215°C to 217°C
Color White
Beilstein 1723527
Solubility Information Soluble in water.
Quantity 1 kg
Formula Weight 150.14 (132.12 Anhydrous)
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Asparagine monohydrate
EINECSNumber 200-735-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only