missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Asparagine monohydrate, 99%

L(+)-Asparagine monohydrate, 99%, C4H10N2O4, CAS Number-5794-13-8, asparagine monohydrate, l +-asparagine monohydrate, asparagine, asparagine nf, l-asparagine hydrate, h-asn-oh.h2o, unii-2pd79vf521, l-asparagine monohydrate, l-asparagine, monohydrate, s-2,4-diamino-4-oxobutanoic acid hydrate, 250g | CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol

$179.17 - $741.61

Chemical Identifiers

CAS 5794-13-8
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
MDL Number MFCD00151038
InChI Key RBMGJIZCEWRQES-DKWTVANSSA-N
Synonym l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521
PubChem CID 170358
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES C(C(C(=O)O)N)C(=O)N.O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC175271000
SDS View Documents
Thermo Scientific Chemicals
175271000
100 g Glass Bottle
Each for $179.17
Only null left
 
AC175272500
SDS View Documents
Thermo Scientific Chemicals
175272500
250 g Glass Bottle
Each for $233.09
Only null left
 
AC175270010
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
175270010
1 kg Glass Bottle
Each for $741.61
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5794-13-8
150.134
RBMGJIZCEWRQES-DKWTVANSSA-N
170358
C(C(C(=O)O)N)C(=O)N.O
C4H10N2O4
MFCD00151038
l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521
(2S)-2,4-diamino-4-oxobutanoic acid;hydrate

Specifications

233.0°C to 235.0°C
5794-13-8
White
10ppm max.
10ppm max.
99%
C4H10N2O4
H2NCOCH2CH(NH2)CO2H·H2O
+ 35.00
98% min. (c=2, H2O, 430nm) 1cm cell
l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521
RBMGJIZCEWRQES-DKWTVANSSA-N
(2S)-2,4-diamino-4-oxobutanoic acid;hydrate
100 g
150.14
Crystalline Powder
1000ppm max.
1ppm max.
210ppm max.
Authentic
11.5% to 12.5% (130°C, 3 hrs)
1% max. (TLC)
Glass Bottle
MFCD00151038
280ppm max.
0.1% max.
Solubility in water: 30g/L water (20°C). Other solubilities: soluble in acids and alkalies, practically insoluble in methanol, ethanol, ether, and benzene
C(C(C(=O)O)N)C(=O)N.O
150.134
170358
99%
L(+)-Asparagine monohydrate, 99%
Videos
SDS
Documents

Documents

RUO – Research Use Only