missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(+)-Asparagine monohydrate, 98+%

CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol

$87.90 - $1267.49

Chemical Identifiers

CAS 5794-13-8
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
MDL Number MFCD00151038
InChI Key RBMGJIZCEWRQES-DKWTVANSSA-N
Synonym l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521
PubChem CID 170358
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES C(C(C(=O)O)N)C(=O)N.O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1501222
View Documents
Thermo Scientific Chemicals
A1501222
100 g
Each for $87.90
Only null left
 
AAA1501236
View Documents
Thermo Scientific Chemicals
A1501236
500 g
Each for $317.75
Only null left
 
AAA150120E
View Documents
Thermo Scientific Chemicals
A150120E
2500 g
Each for $1,267.49
Only null left
 
Description

Description

L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

5794-13-8
150.134
RBMGJIZCEWRQES-DKWTVANSSA-N
170358
C(C(C(=O)O)N)C(=O)N.O
C4H10N2O4
MFCD00151038
l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521
(2S)-2,4-diamino-4-oxobutanoic acid;hydrate

Specifications

∼245°C (decomposition)
C4H10N2O4
5767869
l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521
RBMGJIZCEWRQES-DKWTVANSSA-N
1.543
100 g
170358
150.14 (132.12 Anhydrous)
L-(+)-Asparagine monohydrate
5794-13-8
MFCD00151038
14,837
Soluble in water.
C(C(C(=O)O)N)C(=O)N.O
(2S)-2,4-diamino-4-oxobutanoic acid;hydrate
150.134
+31° (c=5 in 5N HCl)
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-735-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only