missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(+)-Asparagine monohydrate, 98+%

Catalog No. AAA1501236
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1501236 500 g
AAA1501222 100 g
AAA150120E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1501236 Supplier Thermo Scientific Chemicals Supplier No. A1501236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5794-13-8 | C4H10N2O4 | 150.134 g/mol

L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Asparagine is a naturally occurring proteinogenic amino acid, used in biomanufacturing cell culture systems for the production of therapeutic recombinant proteins and monoclonal antibodies. It is used in the biosynthesis of proteins. It is also required for development and function of the brain.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 5794-13-8
Molecular Formula C4H10N2O4
Molecular Weight (g/mol) 150.134
MDL Number MFCD00151038
InChI Key RBMGJIZCEWRQES-DKWTVANSSA-N
Synonym l-asparagine monohydrate, l +-asparagine monohydrate, l-asparagine hydrate, asparagine monohydrate, h-asn-oh.h2o, l-asparagine, monohydrate, asparagine nf, s-2,4-diamino-4-oxobutanoic acid hydrate, asparagine, unii-2pd79vf521
PubChem CID 170358
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid;hydrate
SMILES C(C(C(=O)O)N)C(=O)N.O

Specifications

Melting Point ∼245°C (decomposition)
Beilstein 5767869
Merck Index 14,837
Solubility Information Soluble in water.
Density 1.543
Quantity 500 g
Formula Weight 150.14 (132.12 Anhydrous)
Optical Rotation +31° (c=5 in 5N HCl)
Percent Purity ≥98%
Chemical Name or Material L-(+)-Asparagine monohydrate
EINECSNumber 200-735-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only