missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Ascorbic acid sodium salt, 99%

Catalog No. AC352680050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC352680050 5 g Glass bottle
AC352681000 100 g Plastic bottle
AC352685000 500 g Plastic bottle
AC352680025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC352680050 Supplier Thermo Scientific Chemicals Supplier No. 352680050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 134-03-2 | C6H7NaO6 | 198.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00082340
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
PubChem CID 131674100
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Melting Point 219.0°C to 221.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (Iodimetry)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,819
Specific Rotation + 104.00
Solubility Information (5% in water) Clear colorless to light yellow
Formula Weight 198.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L-Ascorbic acid sodium salt, 99%
Show More Show Less
EINECSNumber 205-126-1
RTECSNumber CI7671000
TSCA TSCA

RUO – Research Use Only