missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic acid sodium salt, 99%

Catalog No. p-7023824
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1775922 100 g
AAA1775936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1775922 Supplier Thermo Scientific Chemicals Supplier No. A1775922
Only null left

CAS: 134-03-2 | C6H10NaO6 | 201.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
L-Ascorbic acid sodium salt is used as an antioxidant, an acidity regulator and an antimicrobial in food stuffs. As a reductant, it is involved in the isolation of chloroplasts. It acts as a coenzyme for many hydroxylation reactions. It plays a vital role to eliminate chronic and acute infections. It finds applications as an additive in curing meat products such as ham and sausage meat. It is widely used as a nutritional supplement in agriculture and animal feed to promote the growth of young animals and prevent scurvy. It is also used as an intermediate in the manufacture of various organic chemicals.

Solubility
Soluble in water. Slightly soluble in alcohol. Insoluble in chloroform and ether.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00082340
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
PubChem CID 131674100
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Melting Point ∼220°C
Quantity 100 g
Beilstein 3767246
Merck Index 14,830
Solubility Information Soluble in water. Slightly soluble in alcohol. Insoluble in chloroform and ether.
Optical Rotation +105° (c=5 in Water)
Formula Weight 198.11
Percent Purity 99%
Chemical Name or Material L-Ascorbic acid sodium salt
EINECSNumber 205-126-1
RTECSNumber CI7671000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only