missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid, ACS Reagent, Reagents

Catalog No. 01336198
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336198 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336198 Supplier Labchem Inc Supplier No. C83014NE100C2

L-Ascorbic Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 50-81-7
Color Colorless/White
Packaging Glass Bottle
Molecular Formula C6H8O6
MDL Number MFCD00064328
Quantity 100 g
Specific Gravity 1.65
Synonym Vitamin C, L-Ascorbic Acid
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one
Molecular Weight (g/mol) 176.12
Grade ACS
Physical Form Solid
Chemical Name or Material L-Ascorbic Acid, ACS Reagent
Show More Show Less