missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid 2-Phosphate Sesquimagnesium Salt Hydrate 98.0+%, TCI America™

$123.03 - $406.37

Chemical Identifiers

CAS 113170-55-1
Molecular Formula C12H18Mg3O18P2
Molecular Weight (g/mol) 585.12
MDL Number MFCD08063372
InChI Key IRBRZNUQJUHOBT-UHFFFAOYNA-N
PubChem CID 129894323
IUPAC Name bis({[5-(1,2-dihydroxyethyl)-4-hydroxy-2-oxo-2,5-dihydrofuran-3-yl]oxy}phosphonic acid) trimagnesium
SMILES [Mg].[Mg].[Mg].OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O.OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A25215G
View Documents
TCI America
A25215G
5 g
Each for $123.03
Only null left
 
A252125G
View Documents
TCI America
A252125G
25 g
Each for $406.37
Only null left
 
Description

Description

Specifications

Chemical Identifiers

113170-55-1
585.12
IRBRZNUQJUHOBT-UHFFFAOYNA-N
bis({[5-(1,2-dihydroxyethyl)-4-hydroxy-2-oxo-2,5-dihydrofuran-3-yl]oxy}phosphonic acid) trimagnesium
C12H18Mg3O18P2
MFCD08063372
129894323
[Mg].[Mg].[Mg].OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O.OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O

Specifications

113170-55-1
MFCD08063372
IRBRZNUQJUHOBT-UHFFFAOYNA-N
bis({[5-(1,2-dihydroxyethyl)-4-hydroxy-2-oxo-2,5-dihydrofuran-3-yl]oxy}phosphonic acid) trimagnesium
129894323
≥98.0% (HPLC)
L-Ascorbic Acid 2-Phosphate Sesquimagnesium Salt Hydrate
C12H18Mg3O18P2
5 g
[Mg].[Mg].[Mg].OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O.OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O
585.12
579.07
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5982

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'