missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603720
Change view
Click to view available options
Quantity:
100 g
1 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603720 100 g Poly Bottle
18603721 1 kg Poly Pail
18603722 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603720 Supplier Spectrum Chemical Mfg Cor Supplier No. A1336100GM

A1336, 74-79-3, C6H14N4O2

Spectrum™ Chemical L-Arginine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Lead (Pb) 5 mg/kg
Loss on Drying 1%
Packaging Poly Bottle
Specific Rotation +26.0° to +27°
Quantity 100 g
Percent Purity 98.5 to 101.5%
Grade FCC