missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine, 99%, MP Biomedicals™

Catalog No. p-4403822
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10073680 100 g
ICN10073690 500 g
ICN10073691 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10073680 Supplier MP Biomedicals Inc Supplier No. 0210073680

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Color White
Assay Percent Range 99%
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Quantity 100 g
Formula Weight 174.2
Percent Purity ≥99%
Grade Cell Culture
Physical Form Powder
Chemical Name or Material L-Arginine free base
EINECSNumber 200-811-1
Recommended Storage Store at room temperature.