missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Arginine, 98+%

CAS: 74-79-3 | C6H14N4O2 | 174.20 g/mol

Supplier:  Thermo Scientific Chemicals 104990025

 View more versions of this product

Catalog No. AC104990025


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(+)-Arginine
200ppm max.
White
10ppm max.
10ppm max.
98+%
Plastic Bottle
MFCD00002635
300ppm max.
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
NC(CCCN=C(N)N)C(O)=O
+27.10° (20°C c=8,6N HCl)
2.5 kg
CHEBI:16467
≥98.0% (on dry substance)
98+%
74-79-3
200ppm max.
Authentic
0.5% max. (1 g, 105°C)
C6H14N4O2
H2NC(=NH)NH(CH2)3CH(NH2)CO2H
+25.8° to +28.5° (20°C, 589nm) (c=8, 6N HCl) on dried substance
0.1% max.
ODKSFYDXXFIFQN-UHFFFAOYNA-N
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
174.20
6322
174.2
Crystalline Powder or Crystals

Chemical Identifiers

74-79-3
174.20
ODKSFYDXXFIFQN-UHFFFAOYNA-N
6322
(2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
C6H14N4O2
MFCD00002635
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
CHEBI:16467
NC(CCCN=C(N)N)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 200-811-1

RTECSNumber : CF1934200

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only