missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Arginine, 98+%

CAS: 74-79-3 | C6H14N4O2 | 174.20 g/mol

Supplier:  Thermo Scientific Chemicals A1573814

 View more versions of this product

Catalog No. AAA1573814


Only null left

Description

Description

L-Arginine is used for heart and blood vessel conditions which includes congestive heart failure (CHF), chest pain, high blood pressure and coronary artery disease. It plays a vital role in the treatment of cardiovascular disease due to it being antiatherogenic, anti-ischemic, antiplatelet and antithrombotic. It acts as a growth stimulant and is involved in the treatment of erectile dysfunction in men. It is an important ingredient of tooth paste which provides effective relief for sensitive teeth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Slightly soluble in ethanol. Insoluble in ethyl ether

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications

Specifications

226°C (decomposition)
74-79-3
C6H14N4O2
1725413
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
ODKSFYDXXFIFQN-UHFFFAOYNA-N
2-amino-5-[(diaminomethylidene)amino]pentanoic acid
25 g
CHEBI:16467
174.2
Air Sensitive
L-Arginine
98+%
MFCD00002635
14,780
Soluble in water. Slightly soluble in ethanol. Insoluble in ethyl ether
NC(CCCN=C(N)N)C(O)=O
174.20
6322
+26° (c=1.6 in 6N HCl)
≥98%

Chemical Identifiers

74-79-3
174.20
ODKSFYDXXFIFQN-UHFFFAOYNA-N
6322
NC(CCCN=C(N)N)C(O)=O
C6H14N4O2
MFCD00002635
l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
CHEBI:16467
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 200-811-1

RTECSNumber : CF1934200

TSCA : Yes

storageNote1 : Air sensitive

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only