missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-alpha-Lecithin, granular, from soybean oil

Catalog No. p-153301
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413102500 250 g Plastic Bottle
AC413100010 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413102500 Supplier Thermo Scientific Chemicals Supplier No. 413102500
Only null left

L-alpha-Lecithin, >-97% (phospholipids) min. (as acetone insoluble), C42H80NO8P, CAS Number-8002-43-5 | CAS: 8002-43-5 | C42H80NO8P | 750 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8002-43-5
Molecular Formula C42H80NO8P
Molecular Weight (g/mol) 750
MDL Number MFCD00082428
InChI Key JLPULHDHAOZNQI-JLOPVYAASA-N
Synonym 3-sn-phosphatidylcholine, 1-hexadecanoyl-2-9e,12e-octadecadienoyl-sn-glycero-3-phosphocholine, 1,2-diacyl-sn-glycero-3-phosphocholine, 3,5,8-trioxa-4-phosphahexacosa-17,20-dien-1-aminium, 4-hydroxy-n,n,n-trimethyl-9-oxo-7-1-oxohexadecyl oxy methyl-, inner salt, 4-oxide, r, l-, a-lecithin, 2-linoleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, a-phosphatidylcholine solution, l-alpha-phosphatidylcholine solution, 2-linoleoyl-1-palmitoyl-sn-glyc-ero-3-phosphocholine
PubChem CID 16213884
ChEBI CHEBI:86658
IUPAC Name [(2R)-3-hexadecanoyloxy-2-[(9E,12E)-octadeca-9,12-dienoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC=CCC=CCCCCC

Specifications

Acidity 35mg KOH/g max.
Color Tan or Yellow
Infrared Spectrum Authentic
Packaging Plastic Bottle
Quantity 250 g
Merck Index 15, 5483
Solubility Information Solubility in water: negligible
Formula Weight 750
Percent Purity ≥97% (phospholipids) min. (as acetone insoluble)
Insoluble Matter 0.02% max. (in hexane)
Physical Form Granules
Chemical Name or Material L-α-Lecithin, Source: Soybean
Show More Show Less
EINECSNumber 232-307-2
RTECSNumber OG7565000
TSCA TSCA
storageNote1 Air sensitive

RUO – Research Use Only