missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Allothreonine 99.0+%, TCI America™
$233.00 - $848.14
Chemical Identifiers
| CAS | 28954-12-3 |
|---|---|
| Molecular Formula | C4H9NO3 |
| Molecular Weight (g/mol) | 119.12 |
| MDL Number | MFCD00064268 |
| InChI Key | AYFVYJQAPQTCCC-HRFVKAFMSA-N |
| Synonym | l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine |
| PubChem CID | 99289 |
| ChEBI | CHEBI:28718 |
| IUPAC Name | (2S,3S)-2-amino-3-hydroxybutanoic acid |
| SMILES | C[C@H](O)[C@H](N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A1519100MG
|
TCI America
A1519100MG |
100 mg |
Each for $233.00
|
|
|||||
|
A15191G
|
TCI America
A15191G |
1 g |
Each for $848.14
|
|
|||||
Chemical Identifiers
| 28954-12-3 | |
| 119.12 | |
| AYFVYJQAPQTCCC-HRFVKAFMSA-N | |
| 99289 | |
| (2S,3S)-2-amino-3-hydroxybutanoic acid |
| C4H9NO3 | |
| MFCD00064268 | |
| l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine | |
| CHEBI:28718 | |
| C[C@H](O)[C@H](N)C(O)=O |
Specifications
| 28954-12-3 | |
| C4H9NO3 | |
| l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine | |
| C[C@H](O)[C@H](N)C(O)=O | |
| 100 mg | |
| 99289 | |
| 119.12 | |
| Crystalline Powder |
| White | |
| MFCD00064268 | |
| AYFVYJQAPQTCCC-HRFVKAFMSA-N | |
| (2S,3S)-2-amino-3-hydroxybutanoic acid | |
| 119.12 | |
| CHEBI:28718 | |
| ≥99.0% (HPLC) | |
| L-Allothreonine |
Safety and Handling
RTECSNumber : BA4055000
TSCA : No