missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Allothreonine 99.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Packaging | Glass Bottle with Plastic Insert |
| Quantity | 1 g |
| Formula Weight | 119.12 |
| Percent Purity | ≥99.0% (HPLC) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Allothreonine |
Safety and Handling
| RTECSNumber | BA4055000 |
| TSCA | No |