missing translation for 'onlineSavingsMsg'
Learn More

L-Alanyl-L-proline 96.0+%, TCI America™

Catalog No. a14955g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14955G Supplier TCI America Supplier No. A14955G

Specifications

Purity ≥98.0% (T)
CAS 13485-59-1
Color White
Format Crystal-Powder at 20°C
Molecular Weight (g/mol) 186.21
Product Type L-Alanyl-L-proline
Quantity 5 g